missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Presenilin-2 C-Terminal Fragment Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Presenilin-2 |
---|---|
Clone | 149903 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Presenilin-2 Monoclonal specifically detects Presenilin-2 in Human samples. It is validated for Western Blot.Specifications
Presenilin-2 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P49810 | |
5664 | |
E. coli-derived recombinant human Presenilin-2 C-Terminal Fragment Glu322-Thr388 Accession # P49810 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Presenilin-2 C-Terminal Fragment in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Presenilin-1 C-Terminal Fragment, rhPresenilin-1 N-Terminal Fragment, or rhPresenilin-2 N-Terminal Fragment is observed. |
149903 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AD3LP, AD4AD3L, AD5, E5-1, EC 3.4.23, EC 3.4.23.-, presenilin 2 (Alzheimer disease 4), presenilin-2, Presenilin2, PS-2, PS2CMD1V, PSEN2, PSNL2, STM2STM-2 | |
PSEN2 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only