missing translation for 'onlineSavingsMsg'
Learn More

Pregnenolone, 99%, Thermo Scientific Chemicals

Catalog Number 10582951
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
145-13-1
C21H32O2
316.47
MFCD00003628
ORNBQBCIOKFOEO-OYZZJKLHSA-N
3-hydroxypregn-5-en-20-one
44307923
1-[(10R,13S)-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone
CC(=O)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
145-13-1
C21H32O2
316.47
MFCD00003628
ORNBQBCIOKFOEO-OYZZJKLHSA-N
3-hydroxypregn-5-en-20-one
44307923
1-[(10R,13S)-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone
CC(=O)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 190°C to 193°C
Color White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 7851
Specific Rotation Condition +28.50° (20°C c=1,EtOH)
Specific Rotation + 28.5
Solubility Information Solubility in water: slightly soluble. Other solubilities: solubility (g/100 mL of solution):,carbon terachloride 0.5 / petroleum ether 0.1 / et,acetone 0.6 / chloroform 17.0 / ethanol 1.9 / benz,isopropanol 1.5,solubility (g/100 mL of solvent):
Formula Weight 316.47
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Pregnenolone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'