missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pregabalin Isopropyl Ester Hydrochloride, Mikromol™
Description
Pregabalin Isopropyl Ester Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Pregabalin Isopropyl Ester Hydrochloride |
| CAS | 1239692-16-0 |
| Analyte or Component Names | Pregabalin Isopropyl Ester Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H23 N O2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | (3S)-3-(Aminomethyl)-5-methylhexanoic acid 1-methylethyl ester hydrochloride (1:1), Pregabalin Isopropyl Ester Hydrochloride |
| InChI Formula | InChI=1S/C11H23NO2.ClH/c1-8(2)5-10(7-12)6-11(13)14-9(3)4;/h8-10H,5-7,12H2,1-4H3;1H/t10-;/m0./s1 |
| SMILES | Cl.CC(C)C[C@H](CN)CC(=O)OC(C)C |
| IUPAC Name | propan-2-yl (3S)-3-(aminomethyl)-5-methylhexanoate;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?