missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint Gobain Life Sciences Pre-drilled Rubber Stopper Without Holes
Use these stoppers for a variety of applications. Saint Gobain Life Sciences Rubber Stoppers Without Holes are ideal for chemical transfer.
Brand: Saint Gobain Life Sciences 201090
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Designed to keep container contents secure
- Ideal for securing glass, metal and plastic containers
- Easy to use
Specifications
Blue | |
36mm | |
34 mm | |
Natural Rubber |
Natural Rubber Stoppers | |
28 mm | |
50/Pk. |