Learn More
PRE-084 hydrochloride, Tocris Bioscience™
Highly selective σ1 agonist
Brand:  Tocris (Bio-Techne) 058910
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13487783
  • £133.65 / 10mg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
High affinity, selective σ1 agonist (Ki values are 2.2 and 13091 nM for σ1 and σ2 receptors respectively). Selective over PCP receptors (IC50 > 100000 nM) and several other receptor systems. Produces antiamnesic and antitussive effects in vivo. Also exhibits protective effects in PC6.3 cells expressing mutant huntingtin by activating NF-κB signaling.
Specifications
Chemical Identifiers
138847-85-5
353.887
2-(4-Morpholinethyl) 1-phenylcyclohexanecarboxylate hydrochloride
C19H28ClNO3
QUJWFJNHTBKCLU-UHFFFAOYSA-N
11314197
Specifications
PRE-084 hydrochloride
>99%
2-(4-Morpholinethyl) 1-phenylcyclohexanecarboxylate hydrochloride
C1CCC(CC1)(C2=CC=CC=C2)C(=O)OCCN3CCOCC3.Cl
353.887
353.89
138847-85-5
C19H28ClNO3
QUJWFJNHTBKCLU-UHFFFAOYSA-N
2-morpholin-4-ylethyl 1-phenylcyclohexane-1-carboxylate;hydrochloride
11314197