missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ PRDX2 Recombinant Human Protein, N-His Tag

Expressed from DNA sequence encoding human PRDX2 (Met 1-Asn 198) fused to polyhistidine tag at N-terminus

Brand:  Sino Biological™ 11255H07B5

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15288455

  • £65.07 / 5µg


Estimated Shipment
08-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Formulation: 50mM Tris, 100mM NaCl, 10% Glycerol, pH 8.0
Specifications

Specifications

Analysis of protein structure and protein-protein interactions
7001
≥90% by SDS-PAGE
Human
<1 EU/μg
PRDX2
Other proteins
PRDX2 Recombinant Human Protein
Lyophilized
24kd
5μg
Full Length, Recombinant
Peroxiredoxin 2
Assay Standard
Insect
His Tag
Videos