Learn More
Sino Biological™ PRC1 Recombinant Human Protein, N-His Tag
Expressed from DNA sequence encoding human PRC1 isoform 1 (Met 1-Ser 620) fused to polyhistidine tag at N-terminus
Brand: Sino Biological™ 13280H07B25
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Expressed from DNA sequence encoding human PRC1 isoform 1 (Met 1-Ser 620) fused to polyhistidine tag at N-terminus
- Formulation: 20mM Tris, 500mM NaCl, 20% Glycerol, 3mM DTT, pH 8.0
Specifications
Protein Structure Analysis, Protein Standard, Cell Biology and Analysis | |
74kd | |
Human | |
<1 EU/μg | |
PRC1 | |
His Tag | |
Other Proteins | |
≥90% by SDS-PAGE |
9055 | |
5 x 5μg | |
Full Length, Recombinant | |
Protein Regulator Of Cytokinesis 1 | |
PRC1 Recombinant Human Protein | |
Insect | |
Lyophilized |
For Research Use Only. Not for use in diagnostic procedures.