missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ PRC1 Recombinant Human Protein, N-His Tag

Expressed from DNA sequence encoding human PRC1 isoform 1 (Met 1-Ser 620) fused to polyhistidine tag at N-terminus

Brand:  Sino Biological™ 13280H07B50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15266305

  • £279.99 / 50µg


Estimated Shipment
09-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Formulation: 20mM Tris, 500mM NaCl, 20% Glycerol, 3mM DTT, pH 8.0
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
9055
≥90% by SDS-PAGE
Human
<1 EU/μg
PRC1
Other Proteins
PRC1 Recombinant Human Protein
Lyophilized
74kd
50μg
Full Length, Recombinant
Protein Regulator Of Cytokinesis 1
Assay Standard
Insect
His Tag
Videos