missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Praseodymium(III) isopropoxide

Brand:  Alfa Aesar™ 039430.03

Code : NEW

Additional Details : CAS Number : 19236-14-7 Weight : 0.00100kg Transport : UN number : 2925 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 11328218

  • £66.89 / 1g


Estimated Shipment date
from Supplier 23-02-2018
Add to basket

Specifications

Chemical Identifiers

Praseodymium(III) isopropoxide
Pr[OHC(CH3)2]3
MFCD00145157
19236-14-7
318.12
UN2925

Specifications

Praseodymium(III) isopropoxide
Powder
318.12
UN2925
175°C (0.04mmHg) (subl.)
19236-14-7
Pr[OHC(CH3)2]3
MFCD00145157
Moisture Sensitive
1g
Videos
SDS