missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) fluoride, anhydrous, 99.9% (metals basis), Thermo Scientific™

Catalog Number p-7051519
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy
13709-46-1
F3Pr
197.90
MFCD00016287
BOTHRHRVFIZTGG-UHFFFAOYSA-K
praseodymium(3+) trifluoride
[F-].[F-].[F-].[Pr+3]
This item is not returnable. View return policy
13709-46-1
F3Pr
197.90
MFCD00016287
BOTHRHRVFIZTGG-UHFFFAOYSA-K
praseodymium(3+) trifluoride
[F-].[F-].[F-].[Pr+3]

For vacuum deposition

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 1395°C
Color Green
Physical Form Pieces
Quantity 10 g
UN Number UN3288
Solubility Information Soluble moderately in strong mineral acids Insoluble in water.
Formula Weight 197.9
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 6.2 g/mL
Assay (metals basis)
Chemical Name or Material Praseodymium(III) fluoride, Anhydrous
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'