missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Praseodymium(III) chloride hydrate, REacton™, 99.99% (REO)

Brand:  Alfa Aesar™ 011238.22

Code : NEW

Additional Details : CAS Number : 19423-77-9 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11326298

  • £134.62 / 100g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

Praseodymium(III) chloride hydrate
Reacton™
MFCD00149828
19423-77-9
247.27 (Anhydrous)
14,7712

Specifications

Praseodymium(III) chloride hydrate
Reacton™
99.99%
247.27 (Anhydrous)
14,7712
115°C
19423-77-9
Wet Crystalline Aggregates
(REO)
MFCD00149828
Odorless
100g
Videos
SDS