missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium foil, 1.0mm (0.04in) thick, 99.5% (REO), Thermo Scientific Chemicals

Catalog Number p-7049308
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
7440-10-0
Pr
140.91
MFCD00011174
PUDIUYLPXJFUGB-UHFFFAOYSA-N
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
23942
CHEBI:49828
praseodymium
[Pr]
This item is not returnable. View return policy
7440-10-0
Pr
140.91
MFCD00011174
PUDIUYLPXJFUGB-UHFFFAOYSA-N
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
23942
CHEBI:49828
praseodymium
[Pr]

Praseodymium is used as an alloying constituent for alloys used to make permanent magnets and flints. Along with neodymium, praseodymium is used to manufacture yellow glass which can be used as eye protection (e.g. for welders).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Color White
Quantity 25 x 25 mm
Solubility Information Insoluble in water.
Percent Purity 99.5%
Odor Odorless
Packaging Mylar under Argon
Weight ≈4.23g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Praseodymium foil, 1.0mm (0.04 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'