missing translation for 'onlineSavingsMsg'
Learn More

Pramipexole dihydrochloride, Tocris Bioscience™

Selective D3 agonist

Brand:  Tocris (Bio-Techne) 417410

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13454284

  • £180.81 / 10mg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Dopamine receptor agonist exhibiting selectivity for the D3 receptor (Ki values are 0.5, 3.3, 3.9 and 3.9 nM for D3, D2S, D2L and D4 receptors respectively). Exhibits negligible affinity for D1 and D5 receptors. Displays activity in the treatment of Parkinson's disease.
Specifications

Chemical Identifiers

104632-25-9
284.243
pramipexole dihydrochloride, mirapex, pramipexole hydrochloride, s-n6-propyl-4,5,6,7-tetrahydrobenzo d thiazole-2,6-diamine dihydrochloride, mirapex er, unii-4r2hd0m28n, s-pramipexole dihydrochloride, bi-sifrol
CHEBI:51148
CCCNC1CCC2=C(C1)SC(=N2)N.Cl.Cl
C10H19Cl2N3S
QMNWXHSYPXQFSK-KLXURFKVSA-N
119569
(6S)-6-N-propyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine;dihydrochloride

Specifications

10mg
104632-25-9
pramipexole dihydrochloride, mirapex, pramipexole hydrochloride, s-n6-propyl-4,5,6,7-tetrahydrobenzo d thiazole-2,6-diamine dihydrochloride, mirapex er, unii-4r2hd0m28n, s-pramipexole dihydrochloride, bi-sifrol
CCCNC1CCC2=C(C1)SC(=N2)N.Cl.Cl
284.243
CHEBI:51148
>99%
Pramipexole dihydrochloride
C10H19Cl2N3S
QMNWXHSYPXQFSK-KLXURFKVSA-N
(6S)-6-N-propyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine;dihydrochloride
119569
284.25
Videos
Documents