missing translation for 'onlineSavingsMsg'
Learn More

PQ 401, Tocris Bioscience™

IGF1R inhibitor

Brand:  Tocris (Bio-Techne) 276810

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13450554

  • £170.52 / 10mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Insulin-like growth factor receptor (IGF1R) inhibitor. Suppresses IGF-stimulated IGF-IR autophosphorylation with an IC50 value of 12 μM. Inhibits growth of MCF-7 breast cancer cells in vitro and in vivo.
Specifications

Chemical Identifiers

196868-63-0
341.795
igf-1r inhibitor ii, 1-5-chloro-2-methoxyphenyl-3-2-methylquinolin-4-yl urea, unii-2n3lv83s8j, n-5-chloro-2-methoxyphenyl-n'-2-methyl-4-quinolinyl urea, n-2-methoxy-5-chlorophenyl-n′-2-methylquinolin-4-yl-urea, d0b4su, urea, n-5-chloro-2-methoxyphenyl-n'-2-methyl-4-quinolinyl, pq hplc , powder, 3-5-chloro-2-methoxyphenyl-1-2-methylquinolin-4-yl urea, n-2-methoxy-5-chlorophenyl-n'-2-methylquinolin-4-yl-urea
1-(5-chloro-2-methoxyphenyl)-3-(2-methylquinolin-4-yl)urea
C18H16ClN3O2
YBLWOZUPHDKFOT-UHFFFAOYSA-N
9549305
CC1=NC2=CC=CC=C2C(=C1)NC(=O)NC3=C(C=CC(=C3)Cl)OC

Specifications

PQ 401
196868-63-0
igf-1r inhibitor ii, 1-5-chloro-2-methoxyphenyl-3-2-methylquinolin-4-yl urea, unii-2n3lv83s8j, n-5-chloro-2-methoxyphenyl-n'-2-methyl-4-quinolinyl urea, n-2-methoxy-5-chlorophenyl-n′-2-methylquinolin-4-yl-urea, d0b4su, urea, n-5-chloro-2-methoxyphenyl-n'-2-methyl-4-quinolinyl, pq hplc , powder, 3-5-chloro-2-methoxyphenyl-1-2-methylquinolin-4-yl urea, n-2-methoxy-5-chlorophenyl-n'-2-methylquinolin-4-yl-urea
CC1=NC2=CC=CC=C2C(=C1)NC(=O)NC3=C(C=CC(=C3)Cl)OC
341.795
341.79
10mg
C18H16ClN3O2
YBLWOZUPHDKFOT-UHFFFAOYSA-N
1-(5-chloro-2-methoxyphenyl)-3-(2-methylquinolin-4-yl)urea
9549305
>99%
Videos
Documents