missing translation for 'onlineSavingsMsg'
Learn More

Cowie Technology™ Polypropylene Stir Bar Retriever

Use to retrieve stir bars from mixing containers. The polypropylene stir bar retriever is chemically resistant, inert, and leak-free. Uses Alnico V magnets at its core.

Brand:  Cowie Technology™ 004.1450

Code : NEW

Additional Details : Weight : 0.03572kg

 View more versions of this product

Product Code. 10496972

  • £4.65 / Each


In Stock 3
Add to basket

Description

Description

  • Inert and leak-free
  • Molded in pure, FDA-approved polypropylene
  • Alnico V magnets
  • Autoclavable
Specifications

Specifications

White
Removal of Stirrer Bars from vessels
1/Ea.
10mm
Polypropylene encapsulated Alnico V magnet core
PTFE Stirrer Bars
Polypropylene Stirrer Bar Retriever
450mm
Videos