missing translation for 'onlineSavingsMsg'
Learn More

PPT, Tocris Bioscience™

Subtype-selective ERα agonist

Brand:  Tocris (Bio-Techne) 1426/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13429483

  • £198.45 / 10mg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, subtype-selective estrogen receptor agonist (EC50 ∽ 200 pM); displays 410-fold selectivity for ERα over ERβ. Prevents ovariectomy-induced weight gain and loss of bone mineral density, and induces gene expression in the hypothalamus following systemic administration in vivo.
Specifications

Chemical Identifiers

263717-53-9
386.451
ppt, 1,3,5-tris 4-hydroxyphenyl-4-propyl-1h-pyrazole, propylpyrazoletriol, propyl pyrazole triol, unii-0t83y6jzpf, 0t83y6jzpf, 4,4',4-4-propyl-1h-pyrazole-1,3,5-triyl triphenol, 1,3,5-tris 4-hydroxyphenyl-4-propylpyrazole, 4,4',4-4-propyl-1h-pyrazole-1,3,5-triyl trisphenol, 4,4',4-4-propylpyrazole-1,3,5-triyl trisphenol
4-[2,3-bis(4-hydroxyphenyl)-4-propyl-1H-pyrazol-5-ylidene]cyclohexa-2,5-dien-1-one
C24H22N2O3
UOSWGERPQQOSHS-UHFFFAOYSA-N
6095481
CCCC1=C(N(NC1=C2C=CC(=O)C=C2)C3=CC=C(C=C3)O)C4=CC=C(C=C4)O

Specifications

10mg
263717-53-9
ppt, 1,3,5-tris 4-hydroxyphenyl-4-propyl-1h-pyrazole, propylpyrazoletriol, propyl pyrazole triol, unii-0t83y6jzpf, 0t83y6jzpf, 4,4',4-4-propyl-1h-pyrazole-1,3,5-triyl triphenol, 1,3,5-tris 4-hydroxyphenyl-4-propylpyrazole, 4,4',4-4-propyl-1h-pyrazole-1,3,5-triyl trisphenol, 4,4',4-4-propylpyrazole-1,3,5-triyl trisphenol
CCCC1=C(N(NC1=C2C=CC(=O)C=C2)C3=CC=C(C=C3)O)C4=CC=C(C=C4)O
386.451
386.45
PPT
C24H22N2O3
UOSWGERPQQOSHS-UHFFFAOYSA-N
4-[2,3-bis(4-hydroxyphenyl)-4-propyl-1H-pyrazol-5-ylidene]cyclohexa-2,5-dien-1-one
6095481
>99%
Videos
Documents