missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP6R3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558405
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ADQDDIGNVS FDRVSDINFT LNTNESGNIA LFEACCKERI QQFDDGGSDE EDIWEEKHIA FTPESQRRSS S Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
PPP6R3 Polyclonal antibody specifically detects PPP6R3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
PPP6R3 | |
Polyclonal | |
Unconjugated | |
PPP6R3 | |
4930528G08Rik; 9130026N02Rik; C11orf23; D19Bwg1430e; D19Ertd703e; DKFZp781E17107; DKFZp781E2374; DKFZp781O2362; FLJ11058; FLJ43065; KIAA1558; MGC125711; MGC125712; mKIAA1558; pluripotent cell-specific 3; Pp6r3; Ppcs3; PPP6R3; Ppp6r3-ps1; Pptcs3; protein phosphatase 6 regulatory subunit 3; protein phosphatase 6, regulatory subunit 3; RGD1307145; SAP190; Sapl; SAPLa; SAPS domain family member 3; SAPS domain family, member 3; SAPS domain family, member 3, pseudogene 1; SAPS3; Saps3-ps1; serine/threonine-protein phosphatase 6 regulatory subunit 3; sporulation-induced transcript 4-associated protein; sporulation-induced transcript 4-associated protein SAPL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55291 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5H9R7 | |
PPP6R3 | |
Recombinant protein corresponding to Human PPP6R3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only