Learn More
PPP6R1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560574
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SALSANDAAL THELLALDVP NTMLDLFFHY VFNNFLHAQV EGCVSTMLSL GPPPDSSPET PIQNPVVKHL LQQCRLVERI LTSWEENDRV QCA Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
PPP6R1 Polyclonal antibody specifically detects PPP6R1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
PPP6R1 | |
Polyclonal | |
Unconjugated | |
Ppp6r1 | |
2010309P17Rik; AI836219; B430201G11Rik; D030074N20Rik; KIAA1115; LOW QUALITY PROTEIN: serine/threonine-protein phosphatase 6 regulatory subunit 1; MGC138185; MGC142003; mKIAA1115; Pp6r1; PPP6R1; protein phosphatase 6 regulatory subunit 1; protein phosphatase 6, regulatory subunit 1; RGD1311409; SAP190; SAPS domain family member 1; SAPS domain family, member 1; Saps1; Serine/threonine-protein phosphatase 6 regulatory subunit 1; Unknown (protein for MGC:128005) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22870 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UPN7 | |
Ppp6r1 | |
Recombinant protein corresponding to Human PPP6R1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only