Learn More
PPP4R4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562170
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SKAQLSQTVQ SRLVSCKILG KLTNKFDAHT IKREILPLVK SLCQDVEYEV RSCMCRQLEN IAQGIGTELT KSVVLPELIE LSRDEGSSVR LAAFET Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 96%.
PPP4R4 Polyclonal antibody specifically detects PPP4R4 in Human samples. It is validated for Immunohistochemistry.Specifications
PPP4R4 | |
Polyclonal | |
Unconjugated | |
PPP4R4 | |
8430415E04Rik; CFAP14; cilia and flagella associated protein 14; HEAT-like repeat-containing protein; KIAA1622; mKIAA1622; PP4R4; PPP4R4; protein phosphatase 4 regulatory subunit 4; protein phosphatase 4, regulatory subunit 4; RGD1560932; Serine/threonine-protein phosphatase 4 regulatory subunit 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57718 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6NUP7 | |
PPP4R4 | |
Recombinant protein corresponding to Human PPP4R4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only