missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R3G Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563295
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DAKKEPGAEC FHFSLCLPPG LQPEDEEDAD ERGVAVHFAV CYRCAQGEYW DNNAGANYTL RY Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 77%.
PPP1R3G Polyclonal antibody specifically detects PPP1R3G in Human samples. It is validated for Immunohistochemistry.Specifications
PPP1R3G | |
Polyclonal | |
Unconjugated | |
PPP1R3G | |
PPP1R3G; Protein phosphatase 1 regulatory subunit 3G; protein phosphatase 1, regulatory (inhibitor) subunit 3G; protein phosphatase 1, regulatory subunit 3G; putative protein phosphatase 1 regulatory inhibitor subunit 3G | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
648791 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
B7ZBB8 | |
PPP1R3G | |
Recombinant protein corresponding to Human PPP1R3G | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only