Learn More
PPP1R26 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560794
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NEKRQHETQK CDGSVEKKPD TNENSAKSLL KSHQEPPTKV VHRQGLLGVQ KEFAFRKPPR LAKMNVQPRS LRSKVTTTQE NEGSTKPATP CRPSEAAQNK G Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 51%.
PPP1R26 Polyclonal antibody specifically detects PPP1R26 in Human samples. It is validated for Immunohistochemistry.Specifications
PPP1R26 | |
Polyclonal | |
Unconjugated | |
PPP1R26 | |
1A6/DRIM (down-regulated in metastasis) interacting protein; DRIM/UTP20 interacting protein; KIAA0649; PPP1R26; Protein phosphatase 1 regulatory subunit 26; protein phosphatase 1, regulatory subunit 26 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9858 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T8A7 | |
PPP1R26 | |
Recombinant protein corresponding to Human PPP1R26 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only