missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPM1E Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557867
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TLAVSRGLGD HQLRVLDTNI QLKPFLLSVP QVTVLDVDQL ELQEDDVVVM ATDG Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 94%.
PPM1E Polyclonal antibody specifically detects PPM1E in Human samples. It is validated for Immunohistochemistry.Specifications
PPM1E | |
Polyclonal | |
Unconjugated | |
PPM1E | |
AW049266; B930008A12Rik; Ca(2+)/calmodulin-dependent protein kinase phosphatase N; calmodulin-dependent protein kinase phosphatase N; caMKN; CaMKP-N; caMKP-nucleus; Kiaa1072; mKIAA1072; nuclear calmodulin-dependent protein kinase phosphatase; partner of PIX 1; partner of PIXA; Partner of PIX-alpha; POPX1; PP2CE; PP2Ceta; PP2C-eta; PP2CH; PPM1E; PPM1M; Protein phosphatase 1E; protein phosphatase 1E (PP2C domain containing); protein phosphatase 1M; protein phosphatase 1M (PP2C domain containing); protein phosphatase 2C eta-2; protein phosphatase 2C isoform eta; protein phosphatase, Mg2+/Mn2+ dependent 1E; protein phosphatase, Mg2+/Mn2+ dependent 1M; protein phosphatase, Mg2+/Mn2+ dependent, 1E; protein phosphatase, Mg2+/Mn2+ dependent, 1M | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
132160, 22843 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WY54, Q96MI6 | |
PPM1E, PPM1M | |
Recombinant protein corresponding to Human PPM1E | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only