missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPM1D Phosphatase Inhibitor, Thermo Scientific™
Acts as a PPM1D inhibitor
£88.20 - £337.00
Specifications
Color | Yellow |
---|---|
Product Type | Phosphatase Inhibitor |
CAS | 176957-55-4 |
Molecular Formula | C15H12OS2 |
Formula Weight | 272.40g/mol |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15496239
|
Thermo Scientific Alfa Aesar
J64165.MA |
10 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15406249
|
Thermo Scientific Alfa Aesar
J64165.MF |
50 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Yellow | |
176957-55-4 | |
272.40g/mol | |
2°C to 8°C | |
Acts as a PPM1D inhibitor |
Phosphatase Inhibitor | |
C15H12OS2 | |
2,5-Bis-(2-Thienylidene)cyclopentanone; CCT007093 | |
Solid |