Learn More
PPM1D Phosphatase Inhibitor, Thermo Scientific™
Acts as a PPM1D inhibitor
Brand: Thermo Scientific Alfa Aesar J64165.MF
Code : NEW
Additional Details : CAS Number : 176957-55-4 Weight : 0.00050kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Yellow | |
176957-55-4 | |
272.40g/mol | |
Hygroscopic | |
Solid | |
50 mg |
Phosphatase Inhibitor | |
C15H12OS2 | |
2,5-Bis-(2-Thienylidene)cyclopentanone; CCT007093 | |
2°C to 8°C | |
Acts as a PPM1D inhibitor |