missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIG Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567601
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Mouse samples.
PPIG Polyclonal antibody specifically detects PPIG in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin).Specifications
PPIG | |
Polyclonal | |
Unconjugated | |
PPIG | |
AU019516; AU022200; B230312B02Rik; CARS-cyclophilin; CARS-Cyp; CASP10; Clk-associating RS-cyclophilin; Cyclophilin G; CYP; Matrin cyclophilin; matrin cyclophilin (matrin-cyp); matrin-cyp; MGC133241; OTTHUMP00000204761; OTTHUMP00000204762; Peptidyl-prolyl cis-trans isomerase G; peptidylprolyl isomerase G; peptidyl-prolyl isomerase G; peptidylprolyl isomerase G (cyclophilin G); peptidyl-prolyl isomerase G (cyclophilin G); PPIase G; PPIG; Rotamase G; SCAF10; SR-cyclophilin; SRCyp; SR-cyp; SR-related CTD-associated factor 10 | |
Rabbit | |
affinity chromatography | |
RUO | |
228005, 9360 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
A2AR02, Q13427 | |
PPIG | |
Fusion protein of human PPIG | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |