missing translation for 'onlineSavingsMsg'
Learn More

PPDA, Tocris Bioscience™

Subtype-selective NR2C/NR2D antagonist

Brand:  Tocris (Bio-Techne) 253050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13461404

  • £557.72 / 50mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Subtype-selective NMDA receptor antagonist that preferentially binds to NR2C/NR2D-containing receptors (Ki values are 0.096, 0.125, 0.31 and 0.55 μM for NR2C, NR2D, NR2B and NR2A subunits respectively).
Specifications

Chemical Identifiers

684283-16-7
378.384
2s,3r-1-phenanthren-2carbonyl piperazine-2,3-dicarboxylic acid, 2s,3r-1-phenanthren-2-ylcarbonyl piperazine-2,3-dicarboxylic acid, 2jl
(2S,3R)-1-(phenanthrene-2-carbonyl)piperazine-2,3-dicarboxylic acid
C21H18N2O5
IWWXIZOMXGOTPP-MSOLQXFVSA-N
20811874
C1CN(C(C(N1)C(=O)O)C(=O)O)C(=O)C2=CC3=C(C=C2)C4=CC=CC=C4C=C3

Specifications

PPDA
684283-16-7
2s,3r-1-phenanthren-2carbonyl piperazine-2,3-dicarboxylic acid, 2s,3r-1-phenanthren-2-ylcarbonyl piperazine-2,3-dicarboxylic acid, 2jl
C1CN(C(C(N1)C(=O)O)C(=O)O)C(=O)C2=CC3=C(C=C2)C4=CC=CC=C4C=C3
378.384
378.38
50mg
C21H18N2O5
IWWXIZOMXGOTPP-MSOLQXFVSA-N
(2S,3R)-1-(phenanthrene-2-carbonyl)piperazine-2,3-dicarboxylic acid
20811874
>98%
Videos