PPD-2 DPD Powder Pop™ Dispensers for Chlorine Testing

Use with any DPD-based photometer or test kit

Overview
Brand: HF Scientific

Manufacturer Part Number: 10447A

UNSPSC: 60104202

Code: 03

Additional Details:
Additional Details: Weight: 1.00000kg



Product Code. 13181491

Quantity Price
1 £ 95.06 / Pack of 2
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Specification

Quantity 400 Tests
For Use With (Application) For free chlorine testing
Product Type Chlorine Reagent Powder Pop Dispenser
For Use With (Equipment) For Hach™ and Hanna photometers
Volume (Metric) 10mL
Documentation