PPD-2 DPD Powder Pop™ Dispensers for Chlorine Testing

Use with any DPD-based photometer or test kit

Overview
Brand: HF Scientific

Manufacturer Part Number: 10439A

UNSPSC: 60104202

Code: 03

Additional Details:
Additional Details: Weight: 1.00000kg



Product Code. 13141501

Quantity Price
1 £ 106.25 / Pack of 2
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Specification

For Use With (Application) For free chlorine testing
For Use With (Equipment) LaMotte™, Orbeco-Hellige, YSI, palintest, tintometer and Orion photometers
Volume (Metric) 10mL
Quantity 400 Tests
Product Type Chlorine Reagent Powder Pop Dispenser
Documentation