PPD-2 DPD Powder Pop™ Dispensers for Chlorine Testing

Use with any DPD-based photometer or test kit

Overview
Brand: HF Scientific

Manufacturer Part Number: 10445A

UNSPSC: 41115311

Code: 03

Additional Details:
Additional Details: Weight: 1.00000kg



Product Code. 13141491

Quantity Price
1 £ 80.73 / Pack of 2
Estimated Shipment
26-05-2017
Add to basket


Description and Specification

Specification

For Use With (Application) For total chlorine testing
For Use With (Equipment) HF scientific and Hach™ photometers
Includes Pocket photometer
Quantity 400 Tests
Volume (Metric) 5mL
Product Type Chlorine Reagent Powder Pop Dispenser
Documentation