missing translation for 'onlineSavingsMsg'
Learn More

(±)-PPCC oxalate, Tocris Bioscience™

Selective sigma (σ) agonist (σ1> σ2)

Brand:  Tocris (Bio-Techne) 387010

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13463734

  • £155.76 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Selective sigma (σ) receptor ligand. Displays high affinity for σ1; also binds at σ2 sites (Ki = 1.5 nM and 50.8 nM respectively). Selective over a range of receptor types including dopaminergic and muscarinic receptors, DAT and SERT.
Specifications

Chemical Identifiers

932736-91-9
939.068
(S™, R™)-2-[(4-Hydroxy-4-phenyl-1-piperidinyl)methyl]-1-(4-methylphenyl)-cyclopropanecarboxylic acid methyl ester
methyl (1S,2R)-2-[(4-hydroxy-4-phenylpiperidin-1-yl)methyl]-1-(4-methylphenyl)cyclopropane-1-carboxylate;methyl (1R,2S)-2-[(4-hydroxy-4-phenylpiperidin-1-yl)methyl]-1-(4-methylphenyl)cyclopropane-1-carboxylate;oxalic acid
C52H62N2O14
IGJDSEGFEVKASG-KLELIYFTSA-N
90488889
CC1=CC=C(C=C1)C2(CC2CN3CCC(CC3)(C4=CC=CC=C4)O)C(=O)OC.CC1=CC=C(C=C1)C2(CC2CN3CCC(CC3)(C4=CC=CC=C4)O)C(=O)OC.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O

Specifications

(±)-PPCC oxalate
932736-91-9
(S™, R™)-2-[(4-Hydroxy-4-phenyl-1-piperidinyl)methyl]-1-(4-methylphenyl)-cyclopropanecarboxylic acid methyl ester
CC1=CC=C(C=C1)C2(CC2CN3CCC(CC3)(C4=CC=CC=C4)O)C(=O)OC.CC1=CC=C(C=C1)C2(CC2CN3CCC(CC3)(C4=CC=CC=C4)O)C(=O)OC.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O
939.068
469.53
10mg
C52H62N2O14
IGJDSEGFEVKASG-KLELIYFTSA-N
methyl (1S,2R)-2-[(4-hydroxy-4-phenylpiperidin-1-yl)methyl]-1-(4-methylphenyl)cyclopropane-1-carboxylate;methyl (1R,2S)-2-[(4-hydroxy-4-phenylpiperidin-1-yl)methyl]-1-(4-methylphenyl)cyclopropane-1-carboxylate;oxalic acid
90488889
>99%
Videos