missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ PPARG Recombinant Human Protein, N-His.GST tag

Expressed from DNA sequence encoding human PPARG isoform 2 (Met 1-Tyr 505) fused to N-terminal polyhistidine-tagged GST tag at N-Terminus

Brand:  Sino Biological™ 12019H20B50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15295685

  • £641.26 / 50µg


Estimated Shipment
10-07-2018
Add to basket

Description

Description

  • Formulation: 50mM Tris, 100mM NaCl, 20% Glycerol, 0.3 mM DTT, pH 7.4
Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
5468
≥90% by SDS-PAGE
Human
<1 EU/μg
PPARG
Other proteins
PPARG Recombinant Human Protein
Lyophilized
85.4kd
50μg
Full Length, Recombinant
Peroxisome Proliferator-Activated Receptor Gamma
Assay Standard
Insect
GST Tag, His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.