Learn More
Abnova™ PPARG/CEN3q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0216.200uL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16003355
  • £2030.00 / 200µL
Estimated Shipment: 04-06-2024
to see stock.
Specifications
Store at 4°C in the dark
Fluorescent In Situ Hybridization (Cell)
CIMT1,NR1C3,PPARG1,PPARG2,PPARgamma
5468
200 μL
Tube
peroxisome proliferator-activated receptor gamma
PPARG
DNA/RNA