missing translation for 'onlineSavingsMsg'
Learn More

PPADS tetrasodium salt, Tocris Bioscience™

Non-selective P2 antagonist

Brand:  Tocris (Bio-Techne) 0625/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13457823

  • £83.79 / 10mg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A non-selective P2 purinergic antagonist. Blocks recombinant P2X1, P2X2, P2X3, P2X5 (IC50 = 1 - 2.6 μM), native P2Y2-like (IC50 ∽ 0.9 mM), and recombinant P2Y4 (IC50 ∽ 15 mM) receptors. Delays onset of calcium responses to mild hypoosmotic stress in cortical slices.

2'5'-isomer also available.

Specifications

Chemical Identifiers

192575-19-2
599.296
ppads sodium salt, 6-2,4-bis sodiooxysulfonyl phenylazo pyridoxal 5'-phosphoric acid disodium salt
tetrasodium;4-[[4-formyl-6-methyl-5-oxido-3-(phosphonatooxymethyl)pyridin-2-yl]diazenyl]-3-sulfobenzenesulfonate
C14H10N3Na4O12PS2
KURWUCJJNVPCHT-UHFFFAOYSA-J
101086948
CC1=C(C(=C(C(=N1)N=NC2=C(C=C(C=C2)S(=O)(=O)[O-])S(=O)(=O)O)COP(=O)([O-])[O-])C=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

10mg
192575-19-2
ppads sodium salt, 6-2,4-bis sodiooxysulfonyl phenylazo pyridoxal 5'-phosphoric acid disodium salt
CC1=C(C(=C(C(=N1)N=NC2=C(C=C(C=C2)S(=O)(=O)[O-])S(=O)(=O)O)COP(=O)([O-])[O-])C=O)[O-].[Na+].[Na+].[Na+].[Na+]
599.296
599.3
PPADS tetrasodium salt
C14H10N3Na4O12PS2
KURWUCJJNVPCHT-UHFFFAOYSA-J
tetrasodium;4-[[4-formyl-6-methyl-5-oxido-3-(phosphonatooxymethyl)pyridin-2-yl]diazenyl]-3-sulfobenzenesulfonate
101086948
>98%
Videos
Documents