missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | PP11 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16878024
|
Proteintech
18002-1-AP-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16868024
|
Proteintech
18002-1-AP-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein with protease activity and is expressed in the placenta. The protein may be useful as a tumor marker. Multiple alternatively spliced transcript variants have been found for this protein.Specifications
PP11 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P21128, Q3V188 | |
19011, 680317, 8909 | |
PP11 Fusion Protein Ag12494 | |
Primary | |
-20°C | |
ENDOU |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
26 serine protease, ENDOU, P11, Placental protein 11, PP11, Protein endoU, PRSS26 | |
Endou | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |