missing translation for 'onlineSavingsMsg'
Learn More

Buchi™ Powder Drying Flasks

Brand:  Buchi™ 041393

Code : 62

Additional Details : Weight : 6.81000kg

Product Code. 12742275

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

For Rotavapor R–250
20L
Flange
150mm
Powder Drying Flask
1Each
Videos