missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium tungsten oxide, 99.5% (metals basis), Thermo Scientific Chemicals
32.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
Potassium tungsten oxide is used in solar energy and water treatment applications. It is used as alkaloids precipitating agent, bronze production and in tungsten refining. It is also used as an analytical reagent. Further, it is added to molten carbonate fuel cell to prevent the corrosion of stainless-steel separator and retards the sintering of the anode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 921°C |
Physical Form | Powder |
Quantity | 25 g |
Merck Index | 14,7694 |
Solubility Information | Soluble in water. |
Formula Weight | 326.04 |
Percent Purity | 99.5% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Density | 3.12 g/mL |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'