missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium tungstate, 99.95%, for trace metal analysis, -100 mesh, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 921°C |
Quantity | 5 g |
Assay Percent Range | 99.95% |
Linear Formula | K2WO4 |
Merck Index | 15,7811 |
Formula Weight | 326.05 |
Percent Purity | 99.95% |
Grade | Trace Metal |
Packaging | Glass bottle |
Chemical Name or Material | Potassium tungstate |
Safety and Handling
Product Identifier
- Potassium tungstate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'