missing translation for 'onlineSavingsMsg'
Learn More

Potassium tungstate, 99.95%, for trace metal analysis, -100 mesh, ACROS Organics™

Brand:  Acros Organics 448330250

Code : NEW

Additional Details : CAS Number : 7790-60-5 Weight : 0.03000kg


Packaging Glass bottle
Quantity 25g

 View more versions of this product

Product Code. 13438688

  • £37.01 / 25g

Estimated Shipment
25-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

7790-60-5
326.05
potassium tungstate, unii-t6vrm0ou6v, t6vrm0ou6v, dipotassium dioxido dioxo tungsten, dipotassium wolframate, potassium tungstate vi, tungstate wo42-, dipotassium, t-4, 2k.wo4, potassium tungstate-w
dipotassium;dioxido(dioxo)tungsten
K2O4W
AAQNGTNRWPXMPB-UHFFFAOYSA-N
3084032
[O-][W](=O)(=O)[O-].[K+].[K+]

Specifications

Potassium tungstate
99.95%
K2WO4
potassium tungstate, unii-t6vrm0ou6v, t6vrm0ou6v, dipotassium dioxido dioxo tungsten, dipotassium wolframate, potassium tungstate vi, tungstate wo42-, dipotassium, t-4, 2k.wo4, potassium tungstate-w
[O-][W](=O)(=O)[O-].[K+].[K+]
326.05
326.05
For trace metal analysis
0.05% max
25g
7790-60-5
K2O4W
15,7811
AAQNGTNRWPXMPB-UHFFFAOYSA-N
dipotassium;dioxido(dioxo)tungsten
3084032
99.95%
Glass bottle
921°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium tungstate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents