missing translation for 'onlineSavingsMsg'
Learn More

Potassium trioxalatoferrate(III) trihydrate, Thermo Scientific Chemicals

Catalog Number 15411187
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
5936-11-8
C6H3FeK3O12
440.22
MFCD00150450
YXTWCYYDGRBUDE-UHFFFAOYSA-K
Potassium iron(III) oxalate
131675837
This item is not returnable. View return policy
5936-11-8
C6H3FeK3O12
440.22
MFCD00150450
YXTWCYYDGRBUDE-UHFFFAOYSA-K
Potassium iron(III) oxalate
131675837

Potassium trioxalatoferrate(III) trihydrate is used in chemical actinometry to measure light flux.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Green
Physical Form Crystalline
Quantity 500 g
UN Number UN3288
Solubility Information Slightly soluble in water.
SMILES C(=O)(C(=O)O)O.C(=O)(C(=O)[O-])O.C(=O)(C(=O)[O-])[O-].[K+].[K+].[K+].[Fe]
IUPAC Name tripotassium;2-hydroxy-2-oxoacetate;iron;oxalate;oxalic acid
Formula Weight 491.26 (437.21 Anhydrous)
Sensitivity Hygroscopic
Density 2.133 g/mL
Chemical Name or Material Potassium trioxalatoferrate(III) trihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'