missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium trioxalatoferrate(III) trihydrate, Thermo Scientific Chemicals
99.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Description
Potassium trioxalatoferrate(III) trihydrate is used in chemical actinometry to measure light flux.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Green |
Physical Form | Crystalline |
Quantity | 100 g |
UN Number | UN3288 |
Solubility Information | Slightly soluble in water. |
SMILES | [K+].[K+].[K+].[Fe].OC(=O)C([O-])=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O |
IUPAC Name | tripotassium iron trihydrogen oxalate |
Formula Weight | 491.26 (437.21 Anhydrous) |
Sensitivity | Hygroscopic |
Density | 2.133 g/mL |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'