missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiocyanate, ACS, 98.5% min, Thermo Scientific Chemicals

Catalog Number p-4407770
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
333-20-0
CKNS
97.176
MFCD00011413
ZNNZYHKDIALBAK-UHFFFAOYSA-M
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
516872
CHEBI:30951
potassium;thiocyanate
C(#N)[S-].[K+]
This item is not returnable. View return policy
333-20-0
CKNS
97.176
MFCD00011413
ZNNZYHKDIALBAK-UHFFFAOYSA-M
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
516872
CHEBI:30951
potassium;thiocyanate
C(#N)[S-].[K+]

Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the convertion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. Also used in moisturizers and oral care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 500°C (decomposition)
Melting Point 172°C to 175°C
Color White
Physical Form Crystals
Quantity 100 g
Assay Percent Range 98.5% min.
Beilstein 3594799
Solubility Information Soluble in water,acetone,and alcohol. When dissolved in its own weight of water,the temperature drops ^=30° Soluble in water,acetone and alcohol.
Formula Weight 97.18
Grade ACS
Odor Odorless
Sensitivity Light and moisture sensitive
Appearance White crystalline
Density 1.89 g/cm3
Chemical Name or Material Potassium thiocyanate, For ACS analysis
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'