missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium thiocyanate, 99+%, for analysis, Thermo Scientific Chemicals
118.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Melting Point | 170.0°C to 179.0°C |
Color | White |
pH | 5.3 to 8.7 (5% soln. at 20°C) |
Physical Form | Crystalline Powder |
Quantity | 2.5 kg |
Assay Percent Range | 99% min. (Argentometry) |
Linear Formula | KSCN |
Fieser | 01,954 |
Merck Index | 15, 7807 |
Solubility Information | Solubility in water: 2170g/L (20°C). Other solubilities: soluble in alcohol and acetone |
Show More |
Safety and Handling
Product Identifier
- Potassium thiocyanate
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- EUH032-Contact with acids liberates very toxic gas.
- P233-Keep container tightly closed.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'