missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiocyanate, 98%, pure, Thermo Scientific Chemicals

Catalog Number p-3523548
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
333-20-0
CKNS
97.18
MFCD00011413
ZNNZYHKDIALBAK-UHFFFAOYSA-M
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
516872
CHEBI:30951
potassium;thiocyanate
C(#N)[S-].[K+]
333-20-0
CKNS
97.18
MFCD00011413
ZNNZYHKDIALBAK-UHFFFAOYSA-M
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
516872
CHEBI:30951
potassium;thiocyanate
C(#N)[S-].[K+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY
Melting Point 170.0°C to 179.0°C
Color Colorless or White
Physical Form Crystals
Quantity 100 g
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula KSCN
Merck Index 15,7807
Solubility Information Solubility in water: 2170g/L (20°C). Other solubilities: soluble in alcohol and acetone
Formula Weight 97.18
Percent Purity 98%
Grade Pure
Packaging Plastic bottle
Specific Gravity 1.886
Density 1.8860g/mL
Chemical Name or Material Potassium thiocyanate
Show More Show Less
compliance-icons
Product Identifier
  • Potassium thiocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P273-Avoid release to the environment.
  • P233-Keep container tightly closed.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'