missing translation for 'onlineSavingsMsg'
Learn More

Potassium thioacetate, 98%, Thermo Scientific Chemicals

Catalog Number p-4510152
20.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
10387-40-3
C2H3KOS
114.2
SDJHDRMYZQFJJO-UHFFFAOYSA-N
potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium
24201352
ethanethioic S-acid;potassium
CC(=O)S.[K]
This item is not returnable. View return policy
10387-40-3
C2H3KOS
114.2
SDJHDRMYZQFJJO-UHFFFAOYSA-N
potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium
24201352
ethanethioic S-acid;potassium
CC(=O)S.[K]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 173.0°C to 176.0°C
Color White to Brown
pH ≤11 (10% aq. soln.)
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 25 g
Assay Percent Range 98%
Linear Formula CH3COSK
Beilstein 02,23
Fieser 01,955
Solubility Information Solubility in water: freely soluble in water
Formula Weight 114.2
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Appearance (10% in water) clear
Chemical Name or Material Potassium thioacetate
Show More Show Less
compliance-icons
Product Identifier
  • Potassium thioacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'