missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetracyanoplatinate(II) trihydrate, 99.9% (metals basis), Pt 44.9% min, Thermo Scientific Chemicals

Catalog Number p-7048438
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
14323-36-5
C4H6K2N4O3Pt
431.40
MFCD00011374
KBALGZKVNFJSRU-UHFFFAOYSA-N
dipotassium tetracyanoplatinumbis(ylium) trihydrate
O.O.O.[K+].[K+].N#C[Pt++](C#N)(C#N)C#N
This item is not returnable. View return policy
14323-36-5
C4H6K2N4O3Pt
431.40
MFCD00011374
KBALGZKVNFJSRU-UHFFFAOYSA-N
dipotassium tetracyanoplatinumbis(ylium) trihydrate
O.O.O.[K+].[K+].N#C[Pt++](C#N)(C#N)C#N

Potassium tetracyanoplatinate(II) trihydrate is used in the preparation other salts like Krogmann's salt, that are highly conducting, one dimensional materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystalline
Quantity 1 g
Linear Formula K2[Pt(CN)4]·3H2O
UN Number UN1588
Merck Index 14,7685
Solubility Information Soluble in hot water
Formula Weight 431.42 (377.37 Anhydrous)
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pt 44.9% min.
Odor Odorless
Assay (metals basis)
Chemical Name or Material Potassium tetracyanoplatinate(II) trihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'