missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium tetracyanopalladate(II) hydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
Description
Potassium tetracyanopalladate(II) hydrate is used as a primary and secondary intermediate in Chemical & pharmaceutical research. it is also used as catalysts, as precursors of metallic palladium, in preparative chemical production, in photography, in electroplating and medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS | 14516-46-2 |
Color | White |
Physical Form | Crystalline |
Quantity | 1 g |
Molecular Formula | C4K2N4Pd |
MDL Number | MFCD00150431 |
Solubility Information | It is slightly soluble in water. It is soluble in organic or non-aqueous solvent. |
InChI Key | NKYOTIKKMTWAOI-UHFFFAOYSA-N |
SMILES | [K+].[K+].N#C[Pd](C#N)(C#N)C#N |
IUPAC Name | dipotassium palladiumtetracarbonitrile |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'