missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrachloroplatinate(II), 99.9% (metals basis), Pt 46.0% min, Thermo Scientific Chemicals

Catalog Number 11301698
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
10025-99-7
Cl4H4K2Pt
419.11
MFCD00011378
JCAXKWJYKUEOFO-UHFFFAOYSA-N
potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate
61440
Cl.Cl.Cl.Cl.[K+].[K+].[Pt--]
This item is not returnable. View return policy
10025-99-7
Cl4H4K2Pt
419.11
MFCD00011378
JCAXKWJYKUEOFO-UHFFFAOYSA-N
potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate
61440
Cl.Cl.Cl.Cl.[K+].[K+].[Pt--]

Potassium tetrachloroplatinate solution is used in the preparation of platinum (Pt) nanoparticles. Ligands like ammonia or triphenyl phosphine can able to replace the chloride ligands to afford various derivatives. The replacement by ammonia results in the preparation of cisplatin, which is used in the cancer treatment. Further, it is used as catalyst in the hydroarylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Color Red to Brown
Physical Form Crystalline
Quantity 1 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in hot water. Slightly soluble in water.
IUPAC Name dipotassium;tetrachloroplatinum(2-)
Formula Weight 415.11
Odor Odorless
Appearance Red-brown crystals
Density 3.38
Chemical Name or Material Potassium tetrachloroplatinate(II), Pt 46.0% min.
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'