missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-pentyloxide, 14-18% w/v in cyclohexane, Thermo Scientific Chemicals

Catalog Number p-7051512
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
41233-93-6
C5H11KO
126.24
MFCD00064808
ZRLVQFQTCMUIRM-UHFFFAOYSA-N
potassium 2-methylbutan-2-olate, potassium tert-amylate, potassium tert-pentoxide, potassium tert-pentylate, potassium tert-pentoxide solution, potassium 2-methyl-2-butoxide, 2-butanol, 2-methyl-, potassium salt, potassium tert-amyloxide, 2-butanol, 2-methyl-, potassium salt 1:1
23683543
[K+].CCC(C)(C)[O-]
This item is not returnable. View return policy
41233-93-6
C5H11KO
126.24
MFCD00064808
ZRLVQFQTCMUIRM-UHFFFAOYSA-N
potassium 2-methylbutan-2-olate, potassium tert-amylate, potassium tert-pentoxide, potassium tert-pentylate, potassium tert-pentoxide solution, potassium 2-methyl-2-butoxide, 2-butanol, 2-methyl-, potassium salt, potassium tert-amyloxide, 2-butanol, 2-methyl-, potassium salt 1:1
23683543
[K+].CCC(C)(C)[O-]

Coreactant for conversion of ketones to oxides

Potassium tert-pentyloxide is used in wide range of applications in organic synthesis, agrochemicals, pharmaceuticals, colorants and aroma chemicals. It is also used in manufacturing detergents and biodiesel. They also act as catalysts in polymerization and isomerizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.80g/mL
Flash Point −18° (0°F)
Linear Formula CH3CH2C(CH3)2OK
Quantity 5 g
UN Number UN2924
Sensitivity Air & Moisture Sensitive
IUPAC Name potassium;2-methylbutan-2-olate
Formula Weight 126.24
Concentration or Composition (by Analyte or Components) 14 to 18% w/v in cyclohexane
Physical Form Liquid
Chemical Name or Material Potassium tert-pentyloxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'