missing translation for 'onlineSavingsMsg'
Learn More

Potassium (tert-butoxymethyl)trifluoroborate, 97%, ACROS Organics™

Brand:  Acros Organics 444840010

Code : 29

Additional Details : CAS Number : 910251-10-4 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 11916081

  • £72.00 / 1g


Estimated Shipment
23-05-2018
Add to basket


Specifications

Chemical Identifiers

Potassium (tert-butoxymethyl)trifluoroborate
97%
C5H11BF3KO
194.05

Specifications

Potassium (tert-butoxymethyl)trifluoroborate
97%
194.05
910251-10-4
C5H11BF3KO
1g
Videos
SDS