missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium (+)-Tartrate, Extra Pure, SLR, Fisher Chemical
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Specifications
CAS | 921-53-9 |
Color | White |
pH | 7.5 to 8.5 |
Physical Form | Solid |
Quantity | 500 g |
Molecular Formula | C4H4K2O6 |
MDL Number | MFCD00013065 |
Synonym | potassium tartrate, dipotassium tartrate, argol, dipotassium 2,3-dihydroxybutanedioate, potassium tatrate, potassium tartarate, ksc492e6f |
InChI Key | AVTYONGGKAJVTE-OLXYHTOASA-L |
SMILES | [K+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'